Merck
  • Determination of 3-methylhistidine and 1-methylhistidine in untreated urine samples by capillary electrophoresis.
Determination of 3-methylhistidine and 1-methylhistidine in untreated urine samples by capillary electrophoresis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-05-19)
Petr Tůma, Eva Samcová, Pavla Balínová
ABSTRACT
Capillary electrophoretic (CE) method was developed for the determination of urinary 3-methylhistidine (3MH) and 1-methylhistidine (1MH) indicating the extent of degradation of skeletal muscle proteins and thereby the state of human health. 3MH, 1MH and histidine can be separated in both acidic and alkaline media, where these amino acids form cation and anion, respectively. The effective mobility of all ionic forms was measured over a broad range of pH (1.67-11.80), which made it possible to evaluate the corresponding dissociation constants. 3MH and 1MH were determined together with creatinine in untreated urine samples with the limit of detection of 2.4 microM (0.4 mgL(-1)) and 3.0 microM (0.5 mgL(-1)), respectively. Determination was fast and took ca. 12 min including the column washing. Method was employed for an analysis of urine collected from healthy individuals, and from the patients hospitalized with obesity and diabetes mellitus II. This analysis has revealed differences between the healthy individuals and the patients pointing to a more extensive degradation of muscle proteins in the latter group.
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%