Merck
  • Induction of CCAAT/enhancer binding protein-delta by cytokinins, but not by retinoic acid, during granulocytic differentiation of human myeloid leukaemia cells.
Induction of CCAAT/enhancer binding protein-delta by cytokinins, but not by retinoic acid, during granulocytic differentiation of human myeloid leukaemia cells.
British journal of haematology (2005-02-03)
Yuki Ishii, Takashi Kasukabe, Yoshio Honma
ABSTRACT
Cytokinins, purine derivatives that act as hormones to control many processes in plants, are very effective at inducing the granulocytic differentiation of human myeloid leukaemia cells. Isopentenyladenine (IPA), a potent cytokinin, significantly induced the expression of CCAAT/enhancer-binding protein (C/EBP)delta, but not C/EBP alpha protein, whereas all-trans retinoic acid, a well-known inducer of granulocytic differentiation, induced C/EBP alpha but not C/EBP delta protein. Antisense oligonucleotide for C/EBP delta, but not C/EBP alpha or C/EBP beta, effectively suppressed IPA-induced differentiation, suggesting that the expression of C/EBP delta protein is necessary for cytokinin-induced differentiation. Although C/EBP alpha is known to be crucial for granulocytic differentiation, the function of C/EBP delta has not been well documented in the regulation of haematopoiesis. The role of C/EBP delta in the granulocytic differentiation of myeloid leukaemia cells is discussed.
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥90%
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, BioReagent, suitable for plant cell culture, 1 mg/mL
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥98.5%