Merck
  • Sialoadhesin, myelin-associated glycoprotein and CD22 define a new family of sialic acid-dependent adhesion molecules of the immunoglobulin superfamily.
Sialoadhesin, myelin-associated glycoprotein and CD22 define a new family of sialic acid-dependent adhesion molecules of the immunoglobulin superfamily.
Current biology : CB (1994-11-01)
S Kelm, A Pelz, R Schauer, M T Filbin, S Tang, M E de Bellard, R L Schnaar, J A Mahoney, A Hartnell, P Bradfield
ABSTRACT
Protein-carbohydrate interactions are believed to be important in many biological processes that involve cell-cell communication. Apart from the selectins, the only well-characterized vertebrate sialic acid-dependent adhesion molecules are CD22 and sialoadhesin; CD22 is a member of the immunoglobulin superfamily that is expressed by B lymphocytes and sialoadhesin is a macrophage receptor. The recent cloning of the gene encoding sialoadhesin has shown that it is also immunoglobulin-like. Both proteins share sequence similarity with the myelin-associated glycoprotein, an adhesion molecule of oligodendrocytes and Schwann cells that has been implicated in the process of myelination, raising the important question of whether myelin-associated glycoprotein is also a sialic acid-binding protein. We have investigated the binding properties of these three receptors when expressed either in monkey COS cells or as chimaeric proteins containing the Fc portion of human immunoglobulin G. We demonstrate that, like sialoadhesin and CD22, myelin-associated glycoprotein mediates cell adhesion by binding to cell-surface glycans that contain sialic acid. We have dissected the specificities of these three adhesins further: whereas sialoadhesin binds equally to the sugar moieties NeuAc alpha 2-->3Gal beta 1-->3(4)GlcNAc or NeuAc alpha 2-->3Gal beta 1-->3GalNAc, myelin-associated glycoprotein recognizes only NeuAc alpha 2-->3Gal beta 1-->3GalNAc and CD22 binds specifically to NeuAc alpha 2-->6Gal beta 1-->4GlcNAc. Furthermore, we show that the recognition of sialylated glycans on the surfaces of particular cell types leads to the selective binding of sialoadhesin to neutrophils, myelin-associated glycoprotein to neurons and CD22 to lymphocytes. Our findings demonstrate that a subgroup of the immunoglobulin superfamily can mediate diverse biological processes through recognition of specific sialylated glycans on cell surfaces. We propose that this subgroup of proteins be called the sialoadhesin family.