Merck
  • Investigation of the silylation of ephedrines using N-methyl-N-trimethylsilyl-trifluoroacetamide.
Investigation of the silylation of ephedrines using N-methyl-N-trimethylsilyl-trifluoroacetamide.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2004-11-04)
Guro Forsdahl, Günter Gmeiner
ABSTRACT
This paper describes a gas chromatography (GC)--mass spectrometry (MS) method for the simultaneous quantification of ephedrine, pseudoephedrine, cathine, norephedrine and methylephedrine in urine. The sample preparation step includes solid phase extraction and derivatisation with N-methyl-N-trimethylsilyl-trifluoroacetamide (MSTFA). An evaluation of various silylation conditions compatible with screening methods in doping control analysis is presented. The method was found to be well suited for quantification of ephedrines in doping control.
Product Description
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥98.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, for silylations, LiChropur