Merck
  • Profiling analysis of oligosaccharides in antibody pharmaceuticals by capillary electrophoresis.
Profiling analysis of oligosaccharides in antibody pharmaceuticals by capillary electrophoresis.
Journal of chromatography. A (2004-10-29)
Satoru Kamoda, Chie Nomura, Mitsuhiro Kinoshita, Saori Nishiura, Rika Ishikawa, Kazuaki Kakehi, Nana Kawasaki, Takao Hayakawa
ABSTRACT
Carbohydrate chains in glycoprotein pharmaceuticals have important roles for the expression of their biological activities. Therefore, development of an assessment method for the carbohydrate chains is an important parameter for quality control of glycoprotein pharmaceuticals such as newly developed therapeutic antibodies. In this report, we applied capillary electrophoresis with laser-induced fluorescence detection to the analysis of carbohydrate chains after releasing with glycoamidase followed by derivatization with 3-aminobenzoic acid. We found that four major oligosaccharides present in antibody pharmaceuticals were successfully separated with good resolution. The present method showed good precision in both migration times and relative peak areas, and gave comparable accuracy with that using a derivatization method with 8-aminopyrene-1,3,6-trisulfonate.
Sigma-Aldrich
3-Aminobenzoic acid, 98%