Merck
  • Rapid and sensitive amino-acid sequencing of cloning Thermus thermophilus HB8 ferredoxin by proteomics.
Rapid and sensitive amino-acid sequencing of cloning Thermus thermophilus HB8 ferredoxin by proteomics.
Journal of proteome research (2004-10-12)
Maki Kaneko, Ryoji Masui, Kojiro Ake, Yukihide Kousumi, Seiki Kuramitsu, Minoru Yamaguchi, Hiroki Kuyama, Eiji Ando, Shigemi Norioka, Takashi Nakazawa, Taka-Aki Okamura, Hitoshi Yamamoto, Norikazu Ueyama
ABSTRACT
Recombinant holo Thermus thermophilus [7Fe-8S] ferredoxin was synthesized by cloning from Thermus thermophilus HB8 gene. A specific sequence (Pro-His-Val-Ile) at the N-terminus of the recombinant ferredoxin was determined by a rapid and highly sensitive mass spectral method using a novel Ru(II) Edman reagent, [(tpy)Ru(tpy-C6H4-NCS)](PF6)2 (tpy=terpyridine). The formation of the recombinant holoTtFd was established by the characteristic absorptions and CD extrema as [7Fe-8S] ferredoxin. The catalytic electron-transfer reactivity of the [7Fe-8S] ferredoxin between ferredoxin-NADP+ reductase and cytochrome c was recognized.
Sigma-Aldrich
2,2′:6′,2′′-Terpyridine, 98%
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, 99.997% trace metals basis
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, ACS reagent, 99%
Sigma-Aldrich
Ammonium iron(II) sulfate hexahydrate, ReagentPlus®, ≥98%