Merck
Delocalized carotenoid cations in relation to the soliton model.
Journal of the American Chemical Society (2004-07-22)
Bjart Frode Lutnaes, Geir Kildahl-Andersen, Jostein Krane, Synnøve Liaaen-Jensen
ABSTRACT
A series of charge-delocalized carotenoid mono- and dications have been prepared by treatment of selected carotenoids with Brønsted and Lewis acids. The detailed structures of the carbocations were established by NMR studies in the temperature range from -10 to -20 degrees C. The general strategy for structure elucidation by NMR of several cationic components in a mixture is outlined. Bond type and regions of bond inversion were established, as well as the charge distribution, which was determined from the difference in (13)C chemical shift at each carbon. This method gave a more accurate estimate for the partial charges than by using the Spiesecke-Schneider relationship. The resulting charge distribution was used as models for the structure of charged solitons. These carotenoid cations have the most delocalized charge so far determined, and the monocations represent the first experimental structure determination of positively charged solitons. The soliton width determined here is in good agreement with the results of previous AM1 calculations.
Sigma-Aldrich
Boron trifluoride diethyl etherate, for synthesis
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis