Merck
  • Reductive dechlorination of tetrachloroethene by a mixed bacterial culture growing on ethyl lactate.
Reductive dechlorination of tetrachloroethene by a mixed bacterial culture growing on ethyl lactate.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2004-07-13)
Jayashree Jayaraj, Karl J Rockne, Randhir S Makkar
ABSTRACT
Chloroethenes like tetrachloroethene (PCE) are the most prevalent groundwater contaminants in the USA. Their presence as nonaqueous phase liquids (NAPLs) makes remediation difficult. Among options for NAPL cleanup, co-solvent injection has demonstrated success. However, the process has the potential to leave considerable residue of the co-solvent as well as residual chloroethene. Our rationale in this study was to examine whether this residual solvent could be a potential electron donor for the remediation of the residual chloroethene. We hypothesized that ethyl lactate, a "green" solvent, could serve both as a NAPL extraction solvent and an electron donor for reductive dechlorination of residual chloroethene. We examined whether a mixed culture known to degrade PCE with lactate could also grow on ethyl lactate and whether it could stimulate PCE dechlorination. Biomass growth and PCE dechlorination were observed by protein and chloride production, respectively, in the culture; with a specific dechlorination rate of 50 150 microg (mg cell d)(-1). Ethyl lactate abiotically breaks down to ethanol and lactate, the latter being a rich source of hydrogen fo reductive dechlorination. The results demonstrate that ethyl lactate may be promising for in situ bioremediation following NAPL extraction.
Sigma-Aldrich
Ethyl lactate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl lactate, ≥98%, FCC, FG
Sigma-Aldrich
(−)-Ethyl L-lactate, 98%
Sigma-Aldrich
(−)-Ethyl L-lactate, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
(−)-Ethyl L-lactate, ≥99.0%