Merck
[Pharmacological study on Veronicastrum sibiricum (L.) Pennell].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (1992-08-01)
B Zhou, X Meng
ABSTRACT
Six compounds were isolated from Veronicastrum sibiricum and identified as D-mannitol, beta-sitosterol, daucosterine, 3-0-acetyloleanolic acid, 3, 4-dimethoxy cinnamic acid and isoferulic acid. Pharmacological experiment has shown that isoferulic acid is anti-inflammatory and analgesic; 3-0-acetyloleanolic acid is anti-inflammatory; mannitol is analgesic, 3, 4-dimethoxy is also anti-inflammatory and analgesic.
Sigma-Aldrich
3,4-Dimethoxycinnamic acid, predominantly trans, 99%