Merck
  • Concentration of fatty acid ethyl esters in hair of alcoholics: comparison to other biological state markers and self reported-ethanol intake.
Concentration of fatty acid ethyl esters in hair of alcoholics: comparison to other biological state markers and self reported-ethanol intake.
Alcohol and alcoholism (Oxford, Oxfordshire) (2003-12-24)
Friedrich Martin Wurst, Stefan Alexson, Manfred Wolfersdorf, Gaby Bechtel, Stephan Forster, Christer Alling, Steina Aradóttir, Katja Jachau, Peter Huber, John P Allen, Volker Auwärter, Fritz Pragst
ABSTRACT
In a variety of clinical and forensic situations long term use of alcohol must be monitored. In this project we explore the utility of fatty acid ethyl esters (FAEE) in this regard. Additionally, we propose a cut-off value of FAEE to distinguish teetotallers/moderate/social drinkers from alcoholics or individuals drinking at harmful levels. FAEE levels from 18 alcohol-dependent patients in detoxification were contrasted with those of 10 social drinkers and 10 teetotallers. FAEE in hair were determined, using headspace solid phase microextraction and gas chromatography mass spectrometry. C(FAEE), as sum of the concentrations of four esters, was compared to a major FAEE, ethyl palmitate. PEth was measured in heparinized whole blood with a high pressure liquid chromatography (HPLC) method. Drinking validation criteria include self reports, phosphatidyl ethanol (PEth) in whole blood as well as the traditional markers of heavy drinking, gamma glutamyl transpeptidase (GGT), mean corpuscular volume (MCV) and carbohydrate deficient transferrin (CDT). Receiver-operating characteristic (ROC) curve analysis for C(FAEE), indicated a sensitivity of 100% and a specificity of 90% for a cut-off of 0.29 ng/mg. By using a cut-off of 0.4 ng/mg, C(FAEE) identified 94.4% correctly. C(FAEE) and ethyl palmitate were significantly associated (r = 0.945; P < 0.001) as were C(FAEE) and PEth (r = 0.527; P = 0.025). No significant correlation was found between C(FAEE) and total grams of ethanol consumed last month, blood-alcohol concentration at admission to the hospital, CDT, MCV, or GGT. Among the serum and blood markers, %CDT identified 47.1%, MCV 38.8% and GGT 72.2% of patients with chronic intake of higher amounts of ethanol correctly, whereas PEth achieved 100% accuracy. The data suggest that C(FAEE) is a potentially valuable marker of chronic intake of high quantities of ethanol. Furthermore, the results indicate that a reasonable and provisional FAEE cut-off to distinguish between social/moderate and heavy drinking/alcoholism in hair is 0.4 ng/mg.
Sigma-Aldrich
Ethyl palmitate, ≥95%, FG
Sigma-Aldrich
Ethyl palmitate, ≥99%
Sigma-Aldrich
Ethyl stearate, ≥99% (capillary GC)
Sigma-Aldrich
Ethyl myristate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl stearate, ≥97%
Sigma-Aldrich
Ethyl myristate, 99% (GC)