MilliporeSigma
  • Branched aliphatic alkanes with quaternary substituted carbon atoms in modern and ancient geologic samples.
Branched aliphatic alkanes with quaternary substituted carbon atoms in modern and ancient geologic samples.
Proceedings of the National Academy of Sciences of the United States of America (2003-10-11)
Fabien Kenig, Dirk-Jan H Simons, David Crich, James P Cowen, Gregory T Ventura, Tatiana Rehbein-Khalily, Todd C Brown, Ken B Anderson
ABSTRACT
A pseudohomologous series of branched aliphatic alkanes with a quaternary substituted carbon atom (BAQCs, specifically 2,2-dimethylalkanes and 3,3- and 5,5-diethylalkanes) were identified in warm (65 degrees C) deep-sea hydrothermal waters and Late Cretaceous black shales. 5,5-Diethylalkanes were also observed in modern and Holocene marine shelf sediments and in shales spanning the last 800 million years of the geological record. The carbon number distribution of BAQCs indicates a biological origin. These compounds were observed but not identified in previous studies of 2.0 billion- to 2.2 billion-year-old metasediments and were commonly misidentified in other sediment samples, indicating that BAQCs are widespread in the geological record. The source organisms of BAQCs are unknown, but their paleobiogeographic distribution suggests that they have an affinity for sulfides and might be nonphotosynthetic sulfide oxidizers.
Sigma-Aldrich
Pentadecane, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pentadecane, ≥98.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Pentadecane, analytical standard