Merck
  • Enantioselective amitriptyline metabolism in patients phenotyped for two cytochrome P450 isozymes.
Enantioselective amitriptyline metabolism in patients phenotyped for two cytochrome P450 isozymes.
Clinical pharmacology and therapeutics (1992-10-01)
U Breyer-Pfaff, B Pfandl, K Nill, E Nusser, C Monney, M Jonzier-Perey, D Baettig, P Baumann
ABSTRACT
In 26 hospitalized patients with depression, a combined pharmacogenetic test with dextromethorphan, a substrate of cytochrome P450IID6, and mephenytoin, the S-form of which is hydroxylated by a P450IIC isozyme, was carried out before amitriptyline therapy. Metabolites were determined in 24-hour urine samples collected on treatment day 8, and the contributions of individual compounds, including the four isomers of 10-hydroxyamitriptyline and 10-hydroxynortriptyline to total excretion were calculated. Formation of (-)-E-10-hydroxyamitriptyline and (-)-E-10-hydroxynortriptyline apparently depends on the activity of cytochrome P450IID6 because negative correlations existed between the log metabolic ratio of dextromethorphan and the relative quantities of these enantiomers. In contrast, correlations were positive for nortriptyline, (+)-E-10-hydroxynortriptyline, (-)-Z-10-hydroxynortriptyline, and (+)-Z-10-hydroxynortriptyline. The mephenytoin hydroxylase seems to participate in side-chain demethylation to the secondary and primary amines, because the log metabolic ratio of mephenytoin correlated negatively with the relative quantity of E-10-hydroxydidesmethylamitriptyline and positively with that of amitriptyline and its N-glucuronide.