Merck
  • [In situ FTIR spectrocopic investigations of 1,3-butanediol oxidation on Sb and S modified Pt electrodes].
[In situ FTIR spectrocopic investigations of 1,3-butanediol oxidation on Sb and S modified Pt electrodes].
Guang pu xue yu guang pu fen xi = Guang pu (2003-08-30)
Q Wu, Y Yang, Z Zhou, X Xiao, S Cehn, S Sun
ABSTRACT
In the present paper, we studied the surface structure effect of Pt electrode modified with antimony and sulfur adatoms towards 1,3-Butanediol oxidation. The results demonstrated that the modification of Sb and S both inhibited the dissociative adsorption of 1,3-Butanediol into CO, which is the main source of self-poisoning in electrocatalysis of small organic molecules. At lower potentials, the principal oxidation pathway of 1,3-Butanediol on Pt/Sbad electrode is towards the production of C=O. However, at higher potentials, the products of CO2 increased. The presence of S(ad) inhibited the oxidation of 1,3-BD at low potentials(< 0.8 V), and yield mainly acid species as products at higher potentials(> 0.8 V).
Sigma-Aldrich
(R)-(−)-1,3-Butanediol, 95%
Sigma-Aldrich
(±)-1,3-Butanediol, ReagentPlus®, 99.5%
Sigma-Aldrich
(±)-1,3-Butanediol, anhydrous, ≥99%