Merck
  • Surface-enhanced Raman spectroscopy study on the structure changes of 4-mercaptopyridine adsorbed on silver substrates and silver colloids.
Surface-enhanced Raman spectroscopy study on the structure changes of 4-mercaptopyridine adsorbed on silver substrates and silver colloids.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-12-13)
Jiawen Hu, Bing Zhao, Weiqing Xu, Bofu Li, Yuguo Fan
ABSTRACT
Surface-enhanced Raman scattering (SERS) of 4-mercaptopyridine (4-mpy) adsorbed on HNO3 etched silver foil, chemically deposited silver films (silver mirror) and silver colloids were measured. The SERS study has revealed that 4-mpy was adsorbed onto the three kinds of silver surfaces by a sulfur-silver bond with the plane of pyridine ring being normal to the silver substrates. The structure of 4-mpy adsorbed on the silver surfaces depends largely on the pH values of environment. When the pH values of the environment are changed, the structure of 4-mpy adsorbed on silver surfaces can easily be altered through a protonation or deprotonation reaction occurring on the N atom of the pyridine ring, and the modified structure shows unique characters on the SERS spectrum. Owing to the remarkable enhancement ability of SERS technique and characteristic spectrum of different species, a monolayer of 4-mpy assembled on a silver mirror holds potential as a H+ sensor for highly sensitive detection of the proton concentration in an aqueous solution.
Product Description
Silver, foil, 10mm disks, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.004mm, 99.97%
Silver, foil, 10mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95+%
Silver, foil, 0.5m coil, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.008mm, 99.95+%
Silver, foil, 10mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 10mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, 0.5m coil, thickness 0.1mm, as rolled, 99.95+%
Silver, foil, 15mm disks, thickness 0.003mm, 99.9%
Silver, foil, 10m coil, thickness 0.0125mm, coil width 9mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.02mm, 99.97%
Silver, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, 10mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, not light tested, 300x300mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.97%
Silver, foil, light tested, 50x50mm, thickness 0.0125mm, 99.95+%
Silver, foil, not light tested, 50x50mm, thickness 0.002mm, 99.97%
Silver, foil, 4mm disks, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 4mm disks, thickness 0.0125mm, 99.95+%
Silver, foil, 50mm disks, thickness 0.05mm, as rolled, 99.95+%
Silver, foil, 4mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 4mm disks, thickness 0.015mm, 99.97%
Silver, foil, 8mm disks, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 8mm disks, thickness 0.008mm, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%