Merck
  • 2,6-Dichloroquinone chlorimide and 7,7,8,8-tetracyanoquinodimethane reagents for the spectrophotometric determination of salbutamol in pure and dosage forms.
2,6-Dichloroquinone chlorimide and 7,7,8,8-tetracyanoquinodimethane reagents for the spectrophotometric determination of salbutamol in pure and dosage forms.
Journal of pharmaceutical and biomedical analysis (2002-06-07)
G G Mohamed, Shaban M Khalil, M A Zayed, M Abd El-Hamid El-Shall
ABSTRACT
A simple, rapid and sensitive spectrophotometric method for the determination of sulbutamol in pure form and in different pharmaceutical preparations has been developed. The charge transfer (CT) reaction between salbutamol as electron donor and 2,6-dichloroquinone chlorimide (DCQ) and 7,7,8,8-tetracyanoquinodimethane (TCNQ) as a pi-electron acceptor have been spectrophotometrically studied. The optimum experimental conditions for these CT reactions have been studied carefully. Beer's law is obeyed over the concentration range of 1.0-30.0 microg ml(-1) and 2.0-20.0 microg ml(-1) for salbutamol using DCQ and TCNQ, respectively. For more accurate results, Ringbom optimum concentration range is calculated and found to be 10.0 to 30.0 and 8.0 to 20 microg ml(-1) for salbutamol using DCQ and TCNQ, respectively. The Sandell sensitivity is found to be 0.011 and 0.010 g cm(-2) for salbutamol using DCQ and TCNQ, respectively, which indicate the high sensitivity of the proposed methods. Relative standard deviations (R.S.D.) of 0.27 to 0.68% and 0.20 to 1.40% (n=5) were obtained for five replicates of salbutamol using DCQ and TCNQ, respectively. The results obtained by the two reagents are comparable with those obtained by British pharmacopoeia assay for the determination of salbutamol in raw materials and in pharmaceutical preparations.