Merck
  • [Severe ocular burns by calcium carbide in a speleologist: a case report].
[Severe ocular burns by calcium carbide in a speleologist: a case report].
Journal francais d'ophtalmologie (2002-04-10)
F Testud, R Voegtlé, J P Nordmann, J Descotes
ABSTRACT
A case of severe ocular burns in an amateur speleologist is reported. The explosion of his acetylene lamp caused the projection of calcium carbide particles, which induced burning of the cornea and conjunctiva in both eyes. He slowly recovered in several months. The pathophysiology of the burns, linked to the in situ production of lime, and their management are discussed.
Sigma-Aldrich
Calcium carbide, pieces, thickness <10 mm , typically, technical grade, ~80%
Sigma-Aldrich
Calcium carbide, granulated, technical, ≥75% (gas-volumetric)