Merck
Three anthrones from Rubus ulmifolius.
Phytochemistry (2002-04-09)
Guido Flamini, Serena Catalano, Chiara Caponi, Lirio Panizzi, Ivano Morelli
ABSTRACT
From the aerial parts of Rubus ulmifolius Schott three new anthrones, rubanthrone A, B and C, have been isolated. Their structures were established by spectral procedures including 1D and 2D NMR techniques and chemical derivatization. Rubanthrone A showed antimicrobial activity against Staphylococcus aureus at 4.5 mg/ml.
Sigma-Aldrich
Anthrone, ACS reagent, 97%