Merck
  • Synthesis of enantiopure non-natural alpha-amino acids using tert-butyl (2S)-2-[bis-(tert-butoxycarbonyl)amino]-5-oxopentanoate as key-intermediate:the first synthesis of(S)-2-amino-oleic acid.
Synthesis of enantiopure non-natural alpha-amino acids using tert-butyl (2S)-2-[bis-(tert-butoxycarbonyl)amino]-5-oxopentanoate as key-intermediate:the first synthesis of(S)-2-amino-oleic acid.
The journal of peptide research : official journal of the American Peptide Society (2001-10-19)
V Constantinou-Kokotou, V Magrioti, T Markidis, G Kokotos
ABSTRACT
A general method for the synthesis of enantiopure non-natural alpha-amino acids is described. The key intermediate tert-butyl (2S)-2-[bis(tert-butoxycarbonyl)amino]-5-oxopentanoate was obtained from l-glutamic acid after suitable protection and selective reduction of the gamma-methyl ester group by DIBALH. Wittig reaction of this chiral aldehyde with various ylides led to a variety of delta,epsilon-unsaturated alpha-amino acids. This methodology was applied to the synthesis of (S)-2-amino-oleic acid.
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in cyclohexane