Merck
  • Positional distribution of highly unsaturated fatty acids in triacyl-sn-glycerols of Artemia nauplii enriched with docosahexaenoic acid ethyl ester.
Positional distribution of highly unsaturated fatty acids in triacyl-sn-glycerols of Artemia nauplii enriched with docosahexaenoic acid ethyl ester.
Lipids (2001-08-28)
Y Ando, Y Oomi
ABSTRACT
This paper presents the positional distribution of fatty acids in triacyl-sn-glycerols (TAG) of Artemia nauplii used in aquaculture as a live food for marine fish larvae. The nauplii were enriched with docosahexaenoic acid (DHA) ethyl ester (EE) in the form of gelatin-acacia microcapsules for 4, 18, and 24 h. TAG of the initial, enriched, and unenriched Artemia nauplii were subjected to stereospecific analysis. A remarkable increase of DHA content in the enriched Artemia TAG confirmed the view that DHA-EE is effectively assimilated and incorporated into the TAG fraction of Artemia nauplii. TAG of the nauplii enriched with 25 mg/L of DHA-EE contained DHA at concentrations of 5.9-6.8, 4.3-6.0, and 14.3-22.3 mol% in the sn-1, sn-2, and sn-3 positions, respectively. When the nauplii were enriched with 100 mg/L of DHA-EE, proportions of DHA in the sn-1, sn-2, and sn-3 positions were 5.2-8.6, 3.9-6.0, and 12.2-25.4 mol%, respectively. In all of the enriched Artemia, DHA was preferentially located in the sn-3 position followed in sequence by the sn-1 and sn-2 positions. The lower content of DHA in the sn-1 and sn-2 positions was consistent with low content of this acid in 1,2-diacyl-sn-glycerophospholipids. When fish larvae are reared on Artemia nauplii enriched with EE-type DHA oil, the larvae feed on DHA esterified in TAG with a positional distribution pattern similar to that of marine mammals (sn-3 > sn-1 > sn-2) rather than that of fish or marine invertebrates (sn-2 > sn-3 > sn-1).
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid ethyl ester, ≥97%
Pricing and availability is not currently available.
Docosahexaenoic acid ethyl ester, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.