Merck
LC/MS confirmation of ionophores in animal feeds.
Journal of AOAC International (2001-06-22)
S B Turnipseed, J E Roybal, A P Pfenning, S A Gonzales, J A Hurlbut, M R Madson
ABSTRACT
A liquid chromatographic/mass spectrometric (LC/MS) electrospray confirmation method has been developed to confirm 4 ionophores (monensin, lasalocid, salinomycin, and narasin) in a variety of animal feeds using a single quadrupole mass spectrometer. The sodium ions of these compounds are dominant in the electrospray mass spectrum. Using optimized "in-source" collision induced dissociation, characteristic fragment ions seen previously using MS/MS can be observed. The drugs were extracted from the feed matrix using hexane-ethyl acetate and isolated using a silica solid-phase extraction cartridge. These ionophores were confirmed in both medicated feeds and nonmedicated feeds fortified with these drugs at the 1-50 ppm level. In addition, this method was used to confirm residues of monensin in a nonmedicated feed that was collected from a feed mill immediately after the production of a similar feed that was medicated with high levels of monensin.
Sigma-Aldrich
Narasin from Streptomyces auriofaciens, ≥98% (HPLC)