Merck
  • Redescription of Psilotricha acuminata Stein, 1859 and revisions of the genera Psilotricha and Urospinula (Ciliophora, Hypotrichida).
Redescription of Psilotricha acuminata Stein, 1859 and revisions of the genera Psilotricha and Urospinula (Ciliophora, Hypotrichida).
The Journal of eukaryotic microbiology (2001-06-20)
G F Esteban, J L Olmo, B J Finlay
ABSTRACT
Psilotricha acuminata was described by Stein in 1859 as the type species of the ciliate genus Psilotricha Stein, 1859. The ciliate has rarely been found since, and its infraciliature has never been described with the aid of silver-impregnation techniques. We have found P. acuminata Stein, 1859 in soil samples from upland grassland in Scotland (U.K.). Living and healthy organisms of P. acuminata are oblong in outline, and dorso-ventrally compressed. They closely resemble ciliates of the genus Euplotes. The main morphological features used for identification of P. acuminata are the very long and sparse cirri, and the two macronuclei. When the ciliate crawls, the cirri appear stiff and directed backwards. Specimens observed from the ventral side have a protruding anterior end, a rounded or acuminate posterior pole, and a "beak-like" projection to the left side of the posterior end. The ciliate shares characters with the Euplotidae (body shape and reduced ciliature) and with the Oxytrichidae (marginal rows, macronuclei, reduced number of transverse cirri). Because the arrangement of the silver-impregnated infraciliature was unknown, and as the only description of the ciliate was that of Stein (1859a, b), the genus Psilotricha became confused with other hypotrich genera, especially Urospinula Corliss, 1960. Here we provide a full redescription of P. acuminata based on living and silver-impregnated specimens, and a revision of the genera Psilotricha Stein, 1859 and Urospinula Corliss, 1960. We resurrect the genus Urospinula, and give an emended diagnosis for both genera. The species now included within the genus Psilotricha are P. acuminata Stein, 1859 (type species); Psilotricha viridis (Penard, 1922) Kahl, 1932; and Psilotricha geleii (Gelei, 1954) Stiller, 1974. Psilotricha viridis sensu Kahl, 1932 and Psilotricha dragescoi Grolière, 1975 are considered incertae sedis.
Sigma-Aldrich
Silver carbonate, 99%
Sigma-Aldrich
Silver carbonate on Celite®, extent of labeling: ~50 wt. % loading
Sigma-Aldrich
Silver carbonate, purum p.a., ≥99.0% (AT)