Merck
Pesticides in precipitation in the Gdańsk region (Poland).
Chemosphere (2001-04-17)
M Grynkiewicz, Z Polkowska, T Górecki, J Namieśnik
ABSTRACT
Selected organonitrogen, organophosphorus and organochlorine pesticides have been determined in precipitation samples collected at 10 sites in the Gdańsk region (northern Poland) over a period of one year (1998). Compounds which were detected most often included simazine (0.11-5.80 ng/l), fenitrothion (0.1-2.10 ng/l), chlorfenvinfos (0.1-1.30 ng/l), gamma-HCH (0.012-5.06 ng/l), heptachlor epoxide (0.05-3.28 ng/l) and aldrin (0.02-3.28 ng/l). The pesticide concentrations in precipitation samples revealed seasonal fluctuations, with higher concentrations observed during the application periods (June and July). The concentrations observed were also affected by the inflow of polluted air masses from the southwest. The total pesticide concentration in the precipitation samples was strongly related to the abundance of green areas in the vicinity of the sampling sites. A weak correlation was also found between the total concentration of organonitrogen and organophosphorus pesticides, and the total concentration of organochlorine pesticides in the samples collected.