Merck
  • Solid-phase syntheses of heterocycles containing the 2-aminothiophenol moiety.
Solid-phase syntheses of heterocycles containing the 2-aminothiophenol moiety.
Journal of combinatorial chemistry (2000-05-29)
T S Yokum, J Alsina, G Barany
ABSTRACT
Efficient and general procedures have been developed for the solid-phase preparation of substituted benzothiazoles (1), 3, 4-dihydro-1,4-benzothiazines (2), 3,4-dihydro-1,4-benzothiazine-1, 1-dioxides (3), 3,4-dihydro-3-oxo-1,4-benzothiazines (4), and 3, 4-dihydro-3-oxo-1,4-benzothiazine-1,1-dioxides (5). All five classes of compounds were prepared from a common intermediate, resin-bound 2-amino-4-carboxythiophenol, in a minimal number of steps. This intermediate was generated by (i) coupling 4-fluoro-3-nitrobenzoic acid onto Wang resin, or onto an amino acid bound to the resin, (ii) substitution of the aryl fluoride with a protected thiol, (iii) reduction of the nitro group, and (iv) removal of sulfur protection. Reaction with the appropriate substrates and reagents to effect cyclization gave the substituted core structures, which were modified further to introduce additional point(s) of diversity. Following cleavages from the solid support, the compounds were obtained in high initial purities and good isolated yields after purification.
Sigma-Aldrich
2-Aminothiophenol, 99%
Sigma-Aldrich
2-Aminothiophenol, technical grade, 90%