Merck
  • Liquid chromatographic determination of vanillin and related aromatic compounds.
Liquid chromatographic determination of vanillin and related aromatic compounds.
Journal of AOAC International (2000-02-29)
E Jagerdeo, E Passetti, S M Dugar
ABSTRACT
This paper describes a reversed-phase liquid chromatographic method for the determination of vanillin, associated natural aromatic compounds and/or synthetic precursors, ethyl vanillin, and coumarin, a commonly encountered adulterant in nonbeverage and beverage alcohol products using a ternary gradient mobile phase. The compounds were separated on a Nova-Pak C18 column by using water, methanol, and tetrahydrofuran as the mobile phase. Measurements were made by using a photodiode array detector at 275 nm. The choice of the mobile phase and the column provides baseline resolution of vanillin and the associated aromatic compounds commonly found in vanilla-flavoring material. Because this method provides low-level detection/quantitation, it is suitable for the characterization of vanilla flavoring materials that are currently added to vanilla flavored beverage alcohol products.
Sigma-Aldrich
Ethyl vanillin, ≥98%, FCC, FG
Sigma-Aldrich
3-Ethoxy-4-hydroxybenzaldehyde, ReagentPlus®, 99%