Merck
  • Arctigenin blocks the unfolded protein response and shows therapeutic antitumor activity.
Arctigenin blocks the unfolded protein response and shows therapeutic antitumor activity.
Journal of cellular physiology (2010-03-17)
Ju-Young Kim, Ji-Hwan Hwang, Mi-Ran Cha, Mi-Young Yoon, Eun-Soon Son, Akihiro Tomida, Bosung Ko, Si-Whan Song, Kazuo Shin-ya, Yong-il Hwang, Hae-Ryong Park
ABSTRACT
Cancer cells in poorly vascularized solid tumors are constantly or intermittently exposed to stressful microenvironments, including glucose deprivation, hypoxia, and other forms of nutrient starvation. These tumor-specific conditions, especially glucose deprivation, activate a signaling pathway called the unfolded protein response (UPR), which enhances cell survival by induction of the stress proteins. We have established a screening method to discover anticancer agents that could preferentially inhibit tumor cell viability under glucose-deprived conditions. Here we identify arctigenin (ARC-G) as an active compound that shows selective cytotoxicity and inhibits the UPR during glucose deprivation. Indeed, ARC-G blocked expression of UPR target genes such as phosphorylated-PERK, ATF4, CHOP, and GRP78, which was accompanied by enhanced phosphorylation of eIF2 alpha during glucose deprivation. The UPR inhibition led to apoptosis involving a mitochondrial pathway by activation of caspase-9 and -3. Furthermore, ARC-G suppressed tumor growth of colon cancer HT-29 xenografts. Our results demonstrate that ARC-G can be served as a novel type of antitumor agent targeting the UPR in glucose-deprived solid tumors.
Sigma-Aldrich
(−)-Arctigenin, ≥95% (LC/MS-ELSD)