Merck
  • Reactions of hydroxyl radical with bergenin, a natural poly phenol studied by pulse radiolysis.
Reactions of hydroxyl radical with bergenin, a natural poly phenol studied by pulse radiolysis.
Bioorganic & medicinal chemistry (2009-07-18)
Umang Singh, Atanu Barik, K Indira Priyadarsini
ABSTRACT
Reactions of pulse radiolytically generated hydroxyl (()OH) radicals and one-electron specific oxidants, Br(2)(-) radicals with bergenin, a polyphenolic tannin derivative, were studied and the transients detected by absorption spectrometry. The transient absorption spectrum produced during the reaction of ()OH radicals with bergenin was broad, and pH dependent. Different modes of reactions of ()OH radicals with bergenin, viz., addition to the aromatic ring adduct and hydrogen abstraction was established by time resolved (5-400micros) transient absorption studies and also by the reaction of Br(2)(-) radicals. Comparing the transient spectra with ()OH radicals and Br(2)(-) radicals at pH 4.5 and 8.5, the absorption maximum of the phenoxyl radical was found to be at 440nm at pH 4.5 and 480nm at pH 8.5. Phenoxyl radicals are produced during ()OH radical reaction through the formation of ()OH radical adduct followed by water/OH(-) elimination. While the phenoxyl radicals of bergenin are oxidizing in nature, the hydroxyl radical adducts and the radicals produced from hydrogen abstraction are of reducing nature. The yield of the oxidizing radicals produced from the ()OH radical reaction with bergenin was determined to be 26.2% by secondary electron transfer reaction from TMPD. On the other hand the yield of reducing radicals produced from the ()OH radical reaction with bergenin was determined to be 74.1% by secondary electron transfer reaction to MV(2+). ()OH radical reactions with bergenin under oxygenated conditions and reaction with trichloro methyl peroxyl radicals with bergenin produced a new transient absorbing at 400nm, which is attributed to peroxyl type of radicals. The one-electron reduction potential for the formation of phenoxyl radical from bergenin was determined to be 0.938V versus NHE at pH 7, by electron transfer equilibrium between bergenin and chlorpromazine. The above results confirmed that reaction of ()OH radicals with bergenin, mainly produced radical adducts and one-electron oxidation accounts to only a minor process. The radical adducts may be converted to peroxyl radicals in presence of oxygen. Based on these results it can be concluded that although bergenin is a polyphenol, it may not act as a potent antioxidant, but may be act as pro-oxidant.
Sigma-Aldrich
Bergenin, ≥95% (LC/MS-ELSD)