Merck
  • The 67-kDa enzymatically inactive alternatively spliced variant of beta-galactosidase is identical to the elastin/laminin-binding protein.
The 67-kDa enzymatically inactive alternatively spliced variant of beta-galactosidase is identical to the elastin/laminin-binding protein.
The Journal of biological chemistry (1998-04-16)
S Privitera, C A Prody, J W Callahan, A Hinek
ABSTRACT
Our previous studies showed immunological and functional similarities, as well as partial sequence homology, between the enzymatically inactive alternatively spliced variant of human beta-galactosidase (S-gal) and the 67-kDa elastin/laminin-binding protein (EBP) from sheep. To define the genetic origin of the EBP further, a full-length human S-gal cDNA clone was constructed and subjected to in vitro transcription/translation. The cDNA was also transfected into COS-1 cells and into the EBP-deficient smooth muscle cells (SMC) from sheep ductus arteriosus (DA). In vitro translation yielded an unglycosylated form of the S-gal protein, which immunoreacted with anti-beta-galactosidase antibodies and bound to elastin and laminin affinity columns. S-gal cDNA transfections into COS-1 and DA SMC increased expression of a 67-kDa protein that immunolocalized intracellularly and to the cell surface and, when extracted from the cells, bound to elastin. The S-gal-transfected cells displayed increased adherence to elastin-covered dishes, consistent with the cell surface distribution of the newly produced S-gal-encoded protein. Transfection of DA SMC additionally corrected their impaired elastic fiber assembly. These results conclusively identify the 67-kDa splice variant of beta-galactosidase as EBP.
Sigma-Aldrich
β-Galactosidase from Aspergillus oryzae, ≥8.0 units/mg solid
Sigma-Aldrich
β-Galactosidase from Escherichia coli, suitable for enzyme immunoassay, lyophilized, powder, ~140 U/mg
Sigma-Aldrich
β-Galactosidase from Escherichia coli, aqueous glycerol suspension, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Galactosidase from bovine testes, ammonium sulfate suspension, 1.0-3.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VI, lyophilized powder, ≥250 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, Grade VIII, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
β-Galactosidase from bovine liver, Grade III, lyophilized powder, ≥0.15 units/mg protein
Sigma-Aldrich
β-Galactosidase from Escherichia coli, lyophilized powder, ≥500 units/mg protein