Merck
  • First morphological characterization of 'Candidatus Mycoplasma turicensis' using electron microscopy.
First morphological characterization of 'Candidatus Mycoplasma turicensis' using electron microscopy.
Veterinary microbiology (2010-12-25)
Barbara Willi, Kristina Museux, Marilisa Novacco, Elisabeth M Schraner, Peter Wild, Katrin Groebel, Urs Ziegler, Godelind A Wolf-Jäckel, Yvonne Kessler, Catrina Geret, Séverine Tasker, Hans Lutz, Regina Hofmann-Lehmann
ABSTRACT
At least three haemotropic mycoplasmas have been recognized in cats: Mycoplasma haemofelis (Mhf), 'Candidatus Mycoplasma haemominutum' (CMhm) and 'Candidatus M. turicensis' (CMt). The latter was originally identified in a Swiss pet cat with haemolytic anaemia and shown to be prevalent in domestic cats and wild felids worldwide using molecular methods. So far, there has been no confirmatory morphological evidence of the existence of CMt presumably due to low blood loads during infection while CMhm has only been characterized by light microscopy with discrepant results. This study aimed to provide for the first time electron microscopic characteristics of CMt and CMhm and to compare them to Mhf. Blood samples from cats experimentally infected with CMt, CMhm and Mhf were used to determine copy numbers in blood by real-time PCR and for transmission and scanning electron microscopy. High resolution scanning electron microscopy revealed CMt and CMhm to be discoid-shaped organisms of 0.3 μm in diameter attached to red blood cells (RBCs). In transmission electron microscopy of CMt, an oval organism of about 0.25 μm with several intracellular electron dense structures was identified close to the surface of a RBC. CMhm and CMt exhibited similar morphology to Mhf but had a smaller diameter. This is the first study to provide morphological evidence of CMt thereby confirming its status as a distinct haemoplasma species, and to present electron microscopic features of CMhm.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dimethyl sulfoxide, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water