Merck
  • Characterization of a novel pyridinium bromide surface confined ionic liquid stationary phase for high-performance liquid chromatography under normal phase conditions via linear solvation energy relationships.
Characterization of a novel pyridinium bromide surface confined ionic liquid stationary phase for high-performance liquid chromatography under normal phase conditions via linear solvation energy relationships.
Journal of chromatography. A (2008-03-07)
D S Van Meter, O D Stuart, A B Carle, A M Stalcup
ABSTRACT
Utilizing linear solvation free energy relationship methodology, a novel pyridinium bromide surface confined ionic liquid (SCIL) stationary phase was characterized under normal phase high-performance liquid chromatographic conditions. A limited set of neutral aromatic probe solutes were utilized to rapidly assess the utility of the LSER model, using mobile phases of hexane modified with 2-propanol. The excellent correlation of the global fit across the mobile phase composition range used in this study for the experimental and calculated retention values (R(2)=0.994) indicates that the LSER model is an appropriate model of characterizing this polar bonded phase under normal phase conditions. For a limited subset of compounds, retention on the pyridinium bromide SCIL stationary phase is more highly correlated with that obtained on a cyano column than on a diol column under NP conditions.
Sigma-Aldrich
2,6-Lutidine
Supelco
2,6-Lutidine, analytical standard
Sigma-Aldrich
2,6-Lutidine, ReagentPlus®, 98%