Merck
  • Combining protein-based IMAC, peptide-based IMAC, and MudPIT for efficient phosphoproteomic analysis.
Combining protein-based IMAC, peptide-based IMAC, and MudPIT for efficient phosphoproteomic analysis.
Journal of proteome research (2008-01-29)
Greg T Cantin, Wei Yi, Bingwen Lu, Sung Kyu Park, Tao Xu, Jiing-Dwan Lee, John R Yates
ABSTRACT
Immobilized metal affinity chromatography (IMAC) is a common strategy used for the enrichment of phosphopeptides from digested protein mixtures. However, this strategy by itself is inefficient when analyzing complex protein mixtures. Here, we assess the effectiveness of using protein-based IMAC as a pre-enrichment step prior to peptide-based IMAC. Ultimately, we couple the two IMAC-based enrichments and MudPIT in a quantitative phosphoproteomic analysis of the epidermal growth factor pathway in mammalian cells identifying 4470 unique phosphopeptides containing 4729 phosphorylation sites.