Merck
The DNA sequence of the human X chromosome.
Nature (2005-03-18)
Mark T Ross, Darren V Grafham, Alison J Coffey, Steven Scherer, Kirsten McLay, Donna Muzny, Matthias Platzer, Gareth R Howell, Christine Burrows, Christine P Bird, Adam Frankish, Frances L Lovell, Kevin L Howe, Jennifer L Ashurst, Robert S Fulton, Ralf Sudbrak, Gaiping Wen, Matthew C Jones, Matthew E Hurles, T Daniel Andrews, Carol E Scott, Stephen Searle, Juliane Ramser, Adam Whittaker, Rebecca Deadman, Nigel P Carter, Sarah E Hunt, Rui Chen, Andrew Cree, Preethi Gunaratne, Paul Havlak, Anne Hodgson, Michael L Metzker, Stephen Richards, Graham Scott, David Steffen, Erica Sodergren, David A Wheeler, Kim C Worley, Rachael Ainscough, Kerrie D Ambrose, M Ali Ansari-Lari, Swaroop Aradhya, Robert I S Ashwell, Anne K Babbage, Claire L Bagguley, Andrea Ballabio, Ruby Banerjee, Gary E Barker, Karen F Barlow, Ian P Barrett, Karen N Bates, David M Beare, Helen Beasley, Oliver Beasley, Alfred Beck, Graeme Bethel, Karin Blechschmidt, Nicola Brady, Sarah Bray-Allen, Anne M Bridgeman, Andrew J Brown, Mary J Brown, David Bonnin, Elspeth A Bruford, Christian Buhay, Paula Burch, Deborah Burford, Joanne Burgess, Wayne Burrill, John Burton, Jackie M Bye, Carol Carder, Laura Carrel, Joseph Chako, Joanne C Chapman, Dean Chavez, Ellson Chen, Guan Chen, Yuan Chen, Zhijian Chen, Craig Chinault, Alfredo Ciccodicola, Sue Y Clark, Graham Clarke, Chris M Clee, Sheila Clegg, Kerstin Clerc-Blankenburg, Karen Clifford, Vicky Cobley, Charlotte G Cole, Jen S Conquer, Nicole Corby, Richard E Connor, Robert David, Joy Davies, Clay Davis, John Davis, Oliver Delgado, Denise Deshazo
ABSTRACT
The human X chromosome has a unique biology that was shaped by its evolution as the sex chromosome shared by males and females. We have determined 99.3% of the euchromatic sequence of the X chromosome. Our analysis illustrates the autosomal origin of the mammalian sex chromosomes, the stepwise process that led to the progressive loss of recombination between X and Y, and the extent of subsequent degradation of the Y chromosome. LINE1 repeat elements cover one-third of the X chromosome, with a distribution that is consistent with their proposed role as way stations in the process of X-chromosome inactivation. We found 1,098 genes in the sequence, of which 99 encode proteins expressed in testis and in various tumour types. A disproportionately high number of mendelian diseases are documented for the X chromosome. Of this number, 168 have been explained by mutations in 113 X-linked genes, which in many cases were characterized with the aid of the DNA sequence.
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Glycerokinase from Bacillus stearothermophilus, buffered aqueous solution, ≥75 units/mg protein (biuret)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Glycerokinase from Escherichia coli, lyophilized powder, 40-100 units/mg protein
Sigma-Aldrich
Glycerokinase from Escherichia coli, 300-600 units/mL, ammonium sulfate suspension
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Monoamine Oxidase A human, recombinant, expressed in baculovirus infected BTI insect cells