Merck
  • Arachidonate lipoxygenase (ALOX) and cyclooxygenase (COX) polymorphisms and colon cancer risk.
Arachidonate lipoxygenase (ALOX) and cyclooxygenase (COX) polymorphisms and colon cancer risk.
Carcinogenesis (2004-08-17)
Julie E Goodman, Elise D Bowman, Stephen J Chanock, Anthony J Alberg, Curtis C Harris
ABSTRACT
In the human colon, arachidonic acid is metabolized primarily by cyclooxygenase (COX) and arachidonate lipoxygenase (ALOX) to bioactive lipids, which are implicated in colon cancer risk. Several polymorphisms in ALOX and COX genes have been identified, including G-1752A, G-1699A and Glu254Lys in ALOX5; Gln261Arg in ALOX12; Leu237Met and Val481Ile in COX1; and C-645T and Val511Ala in COX2. Because of the significant role of arachidonic acid metabolism in colon cancer, we hypothesized that these polymorphisms could influence susceptibility to colon cancer. We addressed this hypothesis in African-Americans and Caucasians using colon cancer cases (n = 293) and hospital- (n = 229) and population-based (n = 304) control groups. Polymorphisms did not differ between the control groups (P > 0.05); thus, they are combined for all analyses presented. ALOX5 Glu254Lys and COX2 C-645T and Val511Ala allele frequencies differed between Caucasians and African-American controls (P < 0.001). The ALOX5 -1752 and -1699 polymorphisms were in linkage disequilibrium (P < 0.001) and associated with a decreased risk in Caucasians in ALOX5 haplotype analyses (P = 0.03). Furthermore, an inverse association was observed between A alleles at positions -1752 and -1699 of ALOX5 and colon cancer risk in Caucasians, but not in African-Americans. Caucasians with A alleles at ALOX5 -1752 had a reduced odds of colon cancer versus those with G alleles [odds ratio (OR) (GA versus GG), 0.63; 95% confidence interval (CI), 0.39-1.01; OR (AA versus GG), 0.33; 95% CI, 0.07-1.65, P(trend) = 0.02]. Similar results were observed for ALOX5 G-1699A [OR (GA versus GG), 0.59, 95% CI, 0.37-0.94; OR (AA versus GG), 0.27, 95% CI, 0.06-1.32, P(trend) = 0.01]. Statistically significant associations with colon cancer were not observed for the other polymorphisms investigated. We have shown for the first time that a haplotype containing ALOX5 G-1752A and G-1699A in a negative regulatory region of the promoter may influence colon cancer risk in Caucasians.
Sigma-Aldrich
Cyclooxygenase 1 from sheep, glycerol solution, ≥1500 units/mg protein
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Please contact Customer Service for Availability
CA$28.30
5 g
Please contact Customer Service for Availability
CA$86.40
25 g
Please contact Customer Service for Availability
CA$247.00