Merck
  • Subcellular localization of multiple PREP2 isoforms is regulated by actin, tubulin, and nuclear export.
Subcellular localization of multiple PREP2 isoforms is regulated by actin, tubulin, and nuclear export.
The Journal of biological chemistry (2004-09-02)
Klaus Haller, Isabel Rambaldi, Eugene Daniels, Mark Featherstone
ABSTRACT
The PREP, MEIS, and PBX families are mammalian members of the TALE (three amino acid loop extension) class of homeodomain-containing transcription factors. These factors have been implicated in cooperative DNA binding with the HOX class of homeoproteins, but PREP and MEIS interact with PBX in apparently non-HOX-dependent cooperative DNA binding as well. PREP, MEIS, and PBX have all been reported to reside in the cytoplasm in one or more tissues of the developing vertebrate embryo. In the case of PBX, cytoplasmic localization is due to the modulation of nuclear localization signals, nuclear export sequences, and interaction with a cytoplasmic anchoring factor, non-muscle myosin heavy chain II B. Here we report that murine PREP2 exists in multiple isoforms distinguished by interaction with affinity-purified antibodies raised to N- and C-terminal epitopes and by nuclear versus cytoplasmic localization. Alternative splicing gives rise to some of these PREP2 isoforms, including a 25-kDa variant lacking the C-terminal half of the protein and homeodomain and having the potential to act as dominant-negative. We further show that cytoplasmic localization is due to the concerted action of nuclear export, as evidenced by sensitivity to leptomycin B, and cytoplasmic retention by the actin and microtubule cytoskeletons. Cytoplasmic PREP2 colocalizes with both the actin and microtubule cytoskeletons and coimmunoprecipitates with actin and tubulin. Importantly, disruption of either cytoskeletal system redirects cytoplasmic PREP2 to the nucleus. We suggest that transcriptional regulation by PREP2 is modulated through the subcellular distribution of multiple isoforms and by interaction with two distinct cytoskeletal systems.
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Phalloidin from Amanita phalloides, ≥90%