Merck
  • Carbapenem resistance due to Bla(OXA-48) among ESBL-producing Escherichia coli and Klebsiella pneumoniae isolates in a univesity hospital, Turkey.
Carbapenem resistance due to Bla(OXA-48) among ESBL-producing Escherichia coli and Klebsiella pneumoniae isolates in a univesity hospital, Turkey.
The Southeast Asian journal of tropical medicine and public health (2013-02-26)
Hasan Nazik, Betigul Ongen, Mehmet Ilktac, Selda Aydin, Nuray Kuvat, Aysegul Sahin, Mucahit Yemisen, Bilgul Mete, Mehmet Sait Durmus, Ilker Inanc Balkan, Ismail Yildiz, Yakup Ergul
ABSTRACT
Bacterial isolates producing Class D OXA-48 carbapenemase may be missed in routine laboratory testing, allowing them to spread undetected. The purpose of the present study was to detect bla(OXA-48) among ESBL-producing Klebsiella pneumoniae and Escherichia coli isolates collected from a university hospital, Turkey. Ninety-two ESBL-producing isolates (66 E. coli, 26 K. pneumoniae) were obtained in 2010. Antibiotic susceptibility tests were performed using the disc diffusion method and VITEK 2 system. Carbapenemase activity was screened using modified Hodge test. Beta-lactamase genes were detected by PCR and bla(OXA-48)-positive amplicons were sequenced. Genetic relatedness among K. pneumoniae isolates was investigated by pulsed-field gel-electrophoresis (PFGE). Carbapenemase activity was detected in 1 E. coli and 9 K. pneumoniae isolates and 8 of the K. pneumoniae plus the E. coli isolates were resistant to ertapenem. Three K. pneumoniae and 1 E. coli isolates were resistant to imipenem. All 10 isolates were susceptible to meropenem. bla(OXA-48) was present in all 10 isolates. Additionally, 9 isolates contained at least one beta-lactamase gene, including bla(SHV') bla(CTX-M) and bla(VEB) type. PFGE revealed different karyotypes among 9 K. pneumoniae isolates suggesting that the dissemination of bla(OXA-48) gene was not spread by a single K. pneumoniae clone. Thus OXA-48-producing isolates found in carbapenem-susceptible strains according to CLSI guidelines.
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)