Merck
  • A novel RING finger gene, SbRFP1, increases resistance to cold-induced sweetening of potato tubers.
A novel RING finger gene, SbRFP1, increases resistance to cold-induced sweetening of potato tubers.
FEBS letters (2013-02-12)
Huiling Zhang, Xun Liu, Jun Liu, Yongbin Ou, Yuan Lin, Meng Li, Botao Song, Conghua Xie
ABSTRACT
The modulation of the activity of enzymes associated with carbohydrate metabolism is important for potato cold-induced sweetening (CIS). A novel RING finger gene SbRFP1 was cloned and its expression was found to be cold-inducible in potato tubers of the CIS-resistant genotypes. Transformation of SbRFP1 in potatoes confirmed its role in inhibiting β-amylase and invertase activity, which consequently slowed down starch and sucrose degradation and the accumulation of reducing sugars in cold stored tubers. These findings strongly suggest that SbRFP1 may function as a negative regulator of BAM1 and StvacINV1 to decelerate the accumulation of reducing sugars in the process of potato CIS.
Product Description
Supelco
Invertase from baker′s yeast (S. cerevisiae), 200-300 units/mg solid
Sigma-Aldrich
Invertase from baker′s yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
Sigma-Aldrich
Invertase from Candida utilis, Grade X, ≥300 units/mg solid
Sigma-Aldrich
β-Amylase from barley, Type II-B, 20-80 units/mg protein (biuret)
Sigma-Aldrich
Invertase Glycoprotein Standard, BioReagent, from Saccharomyces cerevisiae
Sigma-Aldrich
β-Amylase from sweet potato
Sigma-Aldrich
β-Amylase from sweet potato, Type I-B, ammonium sulfate suspension, ≥750 units/mg protein (E1%/280)