Merck
Capillary electrophoretic enzyme assays.
Methods in molecular biology (Clifton, N.J.) (2013-02-07)
Gerhard K E Scriba, Hans Abromeit, Martina Hense, Yi Fan
ABSTRACT
In the past years, capillary electrophoresis has become a frequently used technique for enzyme assays due to the high separation efficiency and versatility as well as small sample size and low consumption of chemicals. The capillary electrophoresis assays can be divided into two general categories: pre-capillary (or offline) assays and in-capillary (or online) assays. In pre-capillary assays, the incubation is performed offline and substrate(s) and product(s) are subsequently analyzed by capillary electrophoresis. In in-capillary assays enzyme reaction and separation of the analytes are performed inside the same capillary. In such assays the enzyme is either immobilized or in solution. The latter techniques is also referred to as electrophoretically mediated microanalysis (EMMA) indicating that the individual steps of the incubation as well as analysis are performed via electrophoretic phenomena. This chapter describes both techniques using the deacetylation of acetyl-lysine residues in model peptides by sirtuin enzymes as well as the hydrolysis of acetylthiocholine by acetylcholinesterase as examples.
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)