Merck
  • Importance of post-treatment follow-up to secure sufficient eradication therapy for Helicobacter pylori.
Importance of post-treatment follow-up to secure sufficient eradication therapy for Helicobacter pylori.
Danish medical journal (2013-01-08)
Stine Roug, Lone Galmstrup Madsen
ABSTRACT
To optimize the care for Helicobacter pylori-associated diseases, we wanted to evaluate the completeness of follow-up after H. pylori eradication therapy in a single Danish endoscopy unit. Furthermore, the eradication rates and possible clinical characteristics associated with failure of eradication therapy were considered. Patients who tested positive for H. pylori infection using a rapid urease test (RUT) during a three-year period were evaluated retrospectively according to demographics, eradication rate, type of eradication therapy, endoscopic findings and number of former attempts of eradication therapy. RUT-positive patients without a post-treatment evaluation were invited for a urea breath test. The overall H. pylori infection rate was 15% (117/796). Only 48/105 (46%) patients had a post-treatment examination to test the effect of H. pylori eradication therapy. The eradication rate by first-line therapy was 75% (58/77). The second-line eradication rate was 87%. 94% (72/77) had the recommended standard triple therapy for first-line eradication therapy. The number of former eradication attempts was the only clinical characteristic that significantly predicted failure of eradication therapy. Among patients with H. pylori-positive peptic ulcer, 21/28 (75%) achieved successful eradication after first-line treatment. Organised follow-up regimes are recommended, especially in patients with absolute treatment indications with a view to optimizing the care for patients infected with H. pylori.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg