Merck
  • The structure of the deacetylase domain of Escherichia coli PgaB, an enzyme required for biofilm formation: a circularly permuted member of the carbohydrate esterase 4 family.
The structure of the deacetylase domain of Escherichia coli PgaB, an enzyme required for biofilm formation: a circularly permuted member of the carbohydrate esterase 4 family.
Acta crystallographica. Section D, Biological crystallography (2013-01-01)
Takashi Nishiyama, Hiroki Noguchi, Hisashi Yoshida, Sam Yong Park, Jeremy R H Tame
ABSTRACT
Bacterial biofilm formation is an extremely widespread phenomenon involving the secretion of a protective exopolysaccharide matrix which helps the bacteria to attach to surfaces and to overcome a variety of stresses in different environments. This matrix may also include proteins, lipids, DNA and metal ions. Its composition depends on the bacterial species and growth conditions, but one of the most widely found components is polymeric β-1,6-N-acetyl-D-glucosamine (PGA). Several studies have suggested that PGA is an essential component of biofilm and it is produced by numerous bacteria, including Escherichia coli, Staphylococcus epidermis, Yersinia pestis, Bordetella spp. and Actinobacillus spp. In E. coli, PGA production and export are dependent on four genes that form a single operon, pgaABCD, which appears to have been transferred between various species. Biofilms themselves are recognized as environments in which such horizontal gene transfer may occur. The pga operon of E. coli, which is even found in innocuous laboratory strains, is highly homologous to that from the plague bacterium Yersinia pestis, and biofilm is believed to play an important role in the transmission of Yersinia. The crystal structure of the N-terminal domain of PgaB, which has deacetylase activity, is described and compared with models of other deacetylases.
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein