Merck
  • Distinct mechanisms regulate ATGL-mediated adipocyte lipolysis by lipid droplet coat proteins.
Distinct mechanisms regulate ATGL-mediated adipocyte lipolysis by lipid droplet coat proteins.
Molecular endocrinology (Baltimore, Md.) (2012-12-04)
Xingyuan Yang, Bradlee L Heckmann, Xiaodong Zhang, Cynthia M Smas, Jun Liu
ABSTRACT
Adipose triglyceride lipase (ATGL) is the key triacylglycerol hydrolase in adipocytes. The precise mechanisms by which ATGL action is regulated by lipid droplet (LD) coat proteins and responds to hormonal stimulation are incompletely defined. By combining usage of loss- and gain-of-function approaches, we sought to determine the respective roles of perilipin 1 and fat-specific protein 27 (FSP27) in the control of ATGL-mediated lipolysis in adipocytes. Knockdown of endogenous perilipin 1 expression resulted in elevated basal lipolysis that was less responsive to β-adrenergic agonist isoproterenol. In comparison, depletion of FSP27 protein increased both basal and stimulated lipolysis with no significant impact on the overall response of cells to isoproterenol. In vitro assays showed that perilipin but not FSP27 was able to inhibit the triacylglycerol hydrolase activity of ATGL. Perilipin 1 also attenuated dose-dependent activation of ATGL by its Coactivator Comparative Gene identification-58. Accordingly, depletion of perilipin 1 and CGI-58 in adipocytes inversely affected basal lipolysis specifically mediated by overexpressed ATGL. Moreover, although depletion of perilipin 1 abolished the LD translocation of ATGL stimulated by isoproterenol, absence of FSP27 resulted in multilocularization of LDs along with increased LD presence of ATGL under both basal and stimulated conditions. Interestingly, knockdown of ATGL expression increased LD size and decreased LD number in FSP27-depeleted cells. Together, our results demonstrate that although FSP27 acts to constitutively limit the LD presence of ATGL, perilipin 1 plays an essential role in mediating the response of ATGL action to β-adrenergic hormones.
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg