Merck
Cu(II) removal from aqueous solution by ureolytic mixed culture (UMC).
Colloids and surfaces. B, Biointerfaces (2012-10-24)
Ismail Simsek, Mustafa Karatas, Emine Basturk
ABSTRACT
The study describes the sorption of living ureolytic mixed culture (UMC) to remove Cu(II) from aqueous solution under various conditions. The effects of various parameters such as optimum biomass, contact time, and Cu(II) concentrations on Cu(II) removal efficiency were investigated. The process was investigated using concentrations of Cu(II) ions ranging from 1 to 200 mg/L in equilibrium batch tests for Langmiur, D-R, and Freundlich isotherm models. The maximum removal efficiency of 99% was obtained a 1 g/L solid-to-liquid ratio with 1-min contact time for 100 mg/L initial concentration of Cu(II).
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, ~1 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), powder, white, ~8 U/mg
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type IX, powder, 50,000-100,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean)
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, glycerol solution, 500-800 units/mL
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid