- Double-knockout of putative endo-β-N-acetylglucosaminidase (ENGase) genes in Arabidopsis thaliana: loss of ENGase activity induced accumulation of high-mannose type free N-glycans bearing N,N'-acetylchitobiosyl unit.
Double-knockout of putative endo-β-N-acetylglucosaminidase (ENGase) genes in Arabidopsis thaliana: loss of ENGase activity induced accumulation of high-mannose type free N-glycans bearing N,N'-acetylchitobiosyl unit.
Bioscience, biotechnology, and biochemistry (2011-05-21)
Yoshinobu Kimura, Yuki Takeoka, Masami Inoue, Megumi Maeda, Kazuhito Fujiyama
PMID21597164
ABSTRACT
Endo-β-N-acetylglucosaminidase (ENGase) is involved in the production of high-mannose type free N-glycans during plant development and fruit maturation. In a previous study (K. Nakamura et al. Biosci. Biotechnol. Biochem., 73, 461-464 (2009)), we identified the tomato ENGase gene and found that gene expression remained relatively constant. In the present study, we constructed an Arabidopsis thaliana mutant in which the expression of two putative ENGase genes was suppressed. The mutant showed no ENGase activity, but produced high-mannose type free N-glycans carrying the N,N'-acetylchitobiosyl unit that is produced by peptide:N-glycanase, indicating that both these genes encode Arabidopsis ENGase.
Sigma-Aldrich
Endoglycosidase F3 from Elizabethkingia miricola, recombinant, expressed in E. coli, 30 U/mg
Sigma-Aldrich
Endoglycosidase F1 from Elizabethkingia miricola, recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F2 from Elizabethkingia miricola, recombinant, expressed in E. coli, 20 U/mg
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution