Merck
  • Some single-nucleotide polymorphisms of the TSSK2 gene may be associated with human spermatogenesis impairment.
Some single-nucleotide polymorphisms of the TSSK2 gene may be associated with human spermatogenesis impairment.
Journal of andrology (2009-11-21)
Hao Zhang, Dan Su, Yuan Yang, Wei Zhang, Yunqiang Liu, Gang Bai, Mingyi Ma, Yongxin Ma, Sizhong Zhang
ABSTRACT
Tssk2, a member of the testis-specific serine/threonine kinase (TSSK) family, is expressed predominantly in the testis and is crucial for the formation and function of sperm cells in the mouse. Targeted deletion of Tssk1 and Tssk2 in male chimeric mice caused infertility because of haploinsufficiency of the genes. Therefore, it is reasonable to postulate that mutations in the human homolog TSSK2 gene may also play a role in impaired spermatogenesis in humans. To explore the possible association between mutations in the TSSK2 gene and idiopathic infertility in humans, mutation screening of the gene in 494 patients with azoospermia or severe oligozoospermia and 357 fertile controls was performed using denaturing high-performance liquid chromatography and DNA sequencing. As a result, 4 single-nucleotide transitions were identified, including c.80A>G (rs3747052), c.774C>T (rs1052756), c.839C>T (rs1052763), and c.1026G>A (rs1052773). Among them, significant differences in polymorphism frequencies were observed of c.80A>G (rs3747052) and c.774C>T (rs1052756) between the patients and controls; the allele G of c.80A>G (rs3747052) and allele T of c.774C>T (rs1052756) seem to be risk factors for the development of spermatogenic impairment, suggesting that the TSSK2 gene may be associated with male idiopathic infertility in humans.
Sigma-Aldrich
Protein Kinase A from bovine heart, ≥0.8 units/μg protein, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Protein Kinase Cζ isozyme human, ≥75% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Protein Kinase Cδ isozyme human, >95% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution
Pricing and availability is not currently available.