Merck
  • Angiopoietin-like 4 protein elevates the prosurvival intracellular O2(-):H2O2 ratio and confers anoikis resistance to tumors.
Angiopoietin-like 4 protein elevates the prosurvival intracellular O2(-):H2O2 ratio and confers anoikis resistance to tumors.
Cancer cell (2011-03-15)
Pengcheng Zhu, Ming Jie Tan, Royston-Luke Huang, Chek Kun Tan, Han Chung Chong, Mintu Pal, Chee Ren Ivan Lam, Petra Boukamp, Jiun Yit Pan, Suat Hoon Tan, Sander Kersten, Hoi Yeung Li, Jeak Ling Ding, Nguan Soon Tan
ABSTRACT
Cancer is a leading cause of death worldwide. Tumor cells exploit various signaling pathways to promote their growth and metastasis. To our knowledge, the role of angiopoietin-like 4 protein (ANGPTL4) in cancer remains undefined. Here, we found that elevated ANGPTL4 expression is widespread in tumors, and its suppression impairs tumor growth associated with enhanced apoptosis. Tumor-derived ANGPTL4 interacts with integrins to stimulate NADPH oxidase-dependent production of O(2)(-). A high ratio of O(2)(-):H(2)O(2) oxidizes/activates Src, triggering the PI3K/PKBα and ERK prosurvival pathways to confer anoikis resistance, thus promoting tumor growth. ANGPTL4 deficiency results in diminished O(2)(-) production and a reduced O(2)(-):H(2)O(2) ratio, creating a cellular environment conducive to apoptosis. ANGPTL4 is an important redox player in cancer and a potential therapeutic target.
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence