Merck
  • Molecular studies of a novel dragline silk from a nursery web spider, Euprosthenops sp. (Pisauridae).
Molecular studies of a novel dragline silk from a nursery web spider, Euprosthenops sp. (Pisauridae).
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2004-08-25)
Natalia N Pouchkina-Stantcheva, Simon J McQueen-Mason
ABSTRACT
Various spider species produce dragline silks with different mechanical properties. The primary structure of silk proteins is thought to contribute to the elasticity and strength of the fibres. Previously published work has demonstrated that the dragline silk of Euprosthenops sp. is stiffer then comparable silk of Nephila edulis, Araneus diadematus and Latrodectus mactans. Our studies of Euprosthenops dragline silk at the molecular level have revealed that nursery web spider fibroin has the highest polyalanine content among previously characterised silks and this is likely to contribute to the superior qualities of pisaurid dragline.