Merck
  • Polyphosphates and pyrophosphates of hexopyranoses as allosteric effectors of human hemoglobin: synthesis, molecular recognition, and effect on oxygen release.
Polyphosphates and pyrophosphates of hexopyranoses as allosteric effectors of human hemoglobin: synthesis, molecular recognition, and effect on oxygen release.
ChemMedChem (2010-11-26)
Konstantina C Fylaktakidou, Carolina D Duarte, Alexandros E Koumbis, Claude Nicolau, Jean-Marie Lehn
ABSTRACT
Polyphosphorylated and perphosphorylated hexopyranose monosaccharides and disaccharides were synthesized from parent or partially protected carbohydrates as potential allosteric effectors of hemoglobin. A study toward the construction of seven- and eight-membered cyclic pyrophosphates was also performed on the sugars which had the proper orientation, protection, and number of phosphates. All final compounds were tested for their efficiency on oxygen release from human hemoglobin. Several compounds presented higher potency than myo-inositol hexakisphosphate, which is the most efficient of the known allosteric effectors of hemoglobin. Structure-activity relationships were analyzed. The affinity and efficiency depend on the number of phosphates attached to the carbohydrate skeleton and are related primarily to the number of negative charges present. Other effects operate, but play a lesser role.
Sigma-Aldrich
AMBERLITE HPR1100, Na ion exchange resin, strongly acidic, 20-50 mesh
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrabutylammonium fluoride hydrate, 98%
Sigma-Aldrich
Tetrazole solution, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Phytase from wheat, ≥0.01 unit/mg solid
Sigma-Aldrich
2,3-Diphospho-D-glyceric acid pentasodium salt, glycolysis metabolite
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium sulfite, anhydrous, Redi-Dri, reagent grade, ≥98%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium sulfite, ≥98%
Supelco
Amberchrom 50WX8, 200-400 mesh, hydrogen form
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl(chloro)diphenylsilane, 98%
Sigma-Aldrich
Palladium on carbon, extent of labeling: 10 wt. % loading, matrix activated carbon support
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dibenzyl N,N-diisopropylphosphoramidite, technical grade, 90%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology