Merck
  • The CXC chemokine SDF-1 is the ligand for LESTR/fusin and prevents infection by T-cell-line-adapted HIV-1.
The CXC chemokine SDF-1 is the ligand for LESTR/fusin and prevents infection by T-cell-line-adapted HIV-1.
Nature (1996-08-29)
E Oberlin, A Amara, F Bachelerie, C Bessia, J L Virelizier, F Arenzana-Seisdedos, O Schwartz, J M Heard, I Clark-Lewis, D F Legler, M Loetscher, M Baggiolini, B Moser
ABSTRACT
A putative chemokine receptor that we previously cloned and termed LESTR has recently been shown to function as a co-receptor (termed fusin) for lymphocyte-tropic HIV-1 strains. Cells expressing CD4 became permissive to infection with T-cell-line-adapted HIV-1 strains of the syncytium-inducing phenotype after transfection with LESTR/fusin complementary DNA. We report here the indentification of a human chemokine of the CXC type, stromal cell-derived factor 1 (SDF-1), as the natural ligand for LESTR/fusin, and we propose the term CXCR-4 for this receptor, in keeping with the new chemokine-receptor nomenclature. SDF-1 activates Chinese hamster ovary (CHO) cells transfected with CXCR-4 cDNA as well as blood leukocytes and lymphocytes. In cell lines expressing CXCR-4 and CD4, and in blood lymphocytes, SDF-1 is a powerful inhibitor of infection by lymphocyte-tropic HIV-1 strains, whereas the CC chemokines RANTES, MIP-1 alpha and MIP-1 beta, which were shown previously to prevent infection with primary, monocyte-tropic viruses, are inactive. In combination with CC chemokines, which block the infection with monocyte/macrophage-tropic viruses, SDF-1 could help to decrease virus load and prevent the emergence of the syncytium-inducing viruses which are characteristic of the late stages of AIDS.
Sigma-Aldrich
Stromal Cell-Derived Factor 1α/pre-B Cell Growth Stimulating Factor human, recombinant, expressed in E. coli, lyophilized powder, ≥97% (SDS-PAGE), suitable for cell culture