Merck
Patterning of antibodies using flexographic printing.
Langmuir : the ACS journal of surfaces and colloids (2012-05-24)
Christopher O Phillips, Sridhar Govindarajan, Simon M Hamblyn, R Steven Conlan, David T Gethin, Tim C Claypole
ABSTRACT
Antibodies were patterned onto flexible plastic films using the flexographic printing process. An ink formulation was developed using high molecular weight polyvinyl alcohol in carbonate-bicarbonate buffer. In order to aid both antibody adhesion and the quality of definition in the printed features, a nitrocellulose coating was developed that was capable of being discretely patterned, thus increasing the signal-to-noise ratio of an antibody array. Printing antibody features such as dots, squares, text, and fine lines were reproduced effectively. Furthermore, this process could be easily adapted for printing of other biological materials, including, but not limited to, enzymes, DNA, proteins, aptamers, and cells.
Sigma-Aldrich
Bicarbonate buffer solution, BioUltra, >0.1 M Na2CO3, >0.2 M NaHCO3
Sigma-Aldrich
Collodion solution, 2% in amyl acetate
Sigma-Aldrich
Collodion solution, 4-8% in ethanol/diethyl ether
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Casein Blocking Buffer 10x, for Northern and Southern blotting, solution
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule