Merck
  • DSCAM and DSCAML1 function in self-avoidance in multiple cell types in the developing mouse retina.
DSCAM and DSCAML1 function in self-avoidance in multiple cell types in the developing mouse retina.
Neuron (2009-12-01)
Peter G Fuerst, Freyja Bruce, Miao Tian, Wei Wei, Justin Elstrott, Marla B Feller, Lynda Erskine, Joshua H Singer, Robert W Burgess
ABSTRACT
DSCAM and DSCAM-LIKE1 (DSCAML1) serve diverse neurodevelopmental functions, including axon guidance, synaptic adhesion, and self-avoidance, depending on the species, cell type, and gene family member studied. We examined the function of DSCAM and DSCAML1 in the developing mouse retina. In addition to a subset of amacrine cells, Dscam was expressed in most retinal ganglion cells (RGCs). RGCs had fasciculated dendrites and clumped cell bodies in Dscam(-/-) mice, suggesting a role in self-avoidance. Dscaml1 was expressed in the rod circuit, and mice lacking Dscaml1 had fasciculated rod bipolar cell dendrites and clumped AII amacrine cell bodies, also indicating a role in self-avoidance. Neurons in Dscam or Dscaml1 mutant retinas stratified their processes appropriately in synaptic laminae in the inner plexiform layer, and functional synapses formed in the rod circuit in mice lacking Dscaml1. Therefore, DSCAM and DSCAML1 function similarly in self-avoidance, and are not essential for synaptic specificity in the mouse retina.
Sigma-Aldrich
Anti-Dihydropyridine-sensitive Calcium Channel α 1 Subunit Antibody, clone 1a, ascites fluid, clone mAB 1a, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ames′ Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Brn-3a Antibody, Chemicon®, from rabbit