Merck
  • Ionizing radiation-dependent and independent phosphorylation of the 32-kDa subunit of replication protein A during mitosis.
Ionizing radiation-dependent and independent phosphorylation of the 32-kDa subunit of replication protein A during mitosis.
Nucleic acids research (2009-08-13)
Holger Stephan, Claire Concannon, Elisabeth Kremmer, Michael P Carty, Heinz-Peter Nasheuer
ABSTRACT
The human single-stranded DNA-binding protein, replication protein A (RPA), is regulated by the N-terminal phosphorylation of its 32-kDa subunit, RPA2. RPA2 is hyperphosphorylated in response to various DNA-damaging agents and also phosphorylated in a cell-cycle-dependent manner during S- and M-phase, primarily at two CDK consensus sites, S23 and S29. Here we generated two monoclonal phospho-specific antibodies directed against these CDK sites. These phospho-specific RPA2-(P)-S23 and RPA2-(P)-S29 antibodies recognized mitotically phosphorylated RPA2 with high specificity. In addition, the RPA2-(P)-S23 antibody recognized the S-phase-specific phosphorylation of RPA2, suggesting that during S-phase only S23 is phosphorylated, whereas during M-phase both CDK sites, S23 and S29, are phosphorylated. Immunofluorescence microscopy revealed that the mitotic phosphorylation of RPA2 starts at the onset of mitosis, and dephosphorylation occurs during late cytokinesis. In mitotic cells treated with ionizing radiation (IR), we observed a rapid hyperphosphorylation of RPA2 in addition to its mitotic phosphorylation at S23 and S29, associated with a significant change in the subcellular localization of RPA. Our data also indicate that the RPA2 hyperphosphorylation in response to IR is facilitated by the activity of both ATM and DNA-PK, and is associated with activation of the Chk2 pathway.
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
G 418 disulfate salt, suitable for plant cell culture, BioReagent
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
G 418 disulfate salt, potency: ≥720 μg per mg (dried basis)
Millipore
ReBlot Plus Kit
Sigma-Aldrich
Roscovitine, A potent, reversible, and selective inhibitor of Cdks that exhibits about 10-fold greater efficacy towards p34-cdk1 and p33-cdk2 and 20-fold greater efficacy towards p33-cdk5 relative to Olomoucine.