Merck
  • A signature of enhanced proliferation associated with response and survival to anti-PD-L1 therapy in early-stage non-small cell lung cancer.
A signature of enhanced proliferation associated with response and survival to anti-PD-L1 therapy in early-stage non-small cell lung cancer.
Cell reports. Medicine (2024-02-25)
Nasser K Altorki, Bhavneet Bhinder, Alain C Borczuk, Olivier Elemento, Vivek Mittal, Timothy E McGraw
ABSTRACT
In early-stage non-small cell lung cancer, the combination of neoadjuvant anti-PD-L1 and subablative stereotactic body radiation therapy (SBRT) is associated with higher rates of major pathologic response compared to anti-PD-L1 alone. Here, we identify a 140-gene set, enriched in genes characteristic of highly proliferating cells, associated with response to the dual therapy. Analysis of on-treatment transcriptome data indicate roles for T and B cells in response. The 140-gene set is associated with disease-free survival when applied to the combined trial arms. This 140-gene set identifies a subclass of tumors in all 7 of The Cancer Genome Atlas tumor types examined. Worse survival is associated with the 140-gene signature in 5 of these tumor types. Collectively, our data support that this 140-gene set, discovered in association with response to combined anti-PD-L1 and SBRT, identifies a clinically aggressive subclass of solid tumors that may be more likely to respond to immunotherapies.